Assay Details
Gene Symbol: TMEM131L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629362.3 | XP_005629419.1 | 32 - 33 | 4374 | 68 | |
XM_022403524.1 | XP_022259232.1 | 30 - 31 | 4168 | 68 | ||
XM_022403525.1 | XP_022259233.1 | 30 - 31 | 4191 | 68 | ||
XM_022403526.1 | XP_022259234.1 | 30 - 31 | 4137 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
