Assay Details
Gene Symbol: SLC50A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003434894.4 | XP_003434942.1 | 2 - 3 | 244 | 58 | |
XM_005622749.3 | XP_005622806.1 | 2 - 3 | 245 | 58 | ||
XM_005622750.3 | XP_005622807.1 | 2 - 3 | 248 | 58 | ||
XM_005622751.3 | XP_005622808.1 | 2 - 3 | 244 | 58 | ||
XM_005622752.3 | XP_005622809.1 | 2 - 3 | 247 | 58 | ||
XM_537256.6 | XP_537256.2 | 2 - 3 | 242 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
