Assay Details
Gene Symbol: PCSK1N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_548989.6 | XP_548989.3 | 2 - 3 | 705 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
