Assay Details
Gene Symbol: PTPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022420800.1 | XP_022276508.1 | 897 | 133 | ||
XM_022420801.1 | XP_022276509.1 | 888 | 133 | |||
XM_843840.5 | XP_848933.1 | 793 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
