Assay Details
Gene Symbol: CNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627618.3 | XP_005627675.1 | 2 | 530 | 261 | |
XM_005627619.3 | XP_005627676.1 | 3 | 613 | 261 | ||
XM_005627620.3 | XP_005627677.1 | 2 | 4559 | 261 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
