Assay Details
| Gene Symbol: SV2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022416774.1 | XP_022272482.1 | 1623 | 152 | ||
| XM_546060.5 | XP_546060.2 | 1851 | 152 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SV2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022416774.1 | XP_022272482.1 | 1623 | 152 | ||
| XM_546060.5 | XP_546060.2 | 1851 | 152 | |||
Set Membership: |
|