Assay Details
Gene Symbol: KCNJ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003120.3 | NP_001003120.1 | 1475 | 97 | ||
GenBank mRNA | AF277647.1 | - | 1137 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
