Assay Details
Gene Symbol: Gngt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496954.1 | XP_003497002.1 | 1 - 2 | 101 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
