Assay Details
| Gene Symbol: Prodh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505568.1 | XP_003505616.1 | 4 - 5 | 599 | 51 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prodh2| Gene Symbol: Prodh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505568.1 | XP_003505616.1 | 4 - 5 | 599 | 51 | |
Set Membership: |
|