Assay Details
Gene Symbol: Npffr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499903.1 | XP_003499951.1 | 1 - 2 | 47 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
