Assay Details
Gene Symbol: Cradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506357.2 | XP_003506405.1 | 2 - 3 | 461 | 70 | |
XM_007646871.1 | XP_007645061.1 | 2 - 3 | 457 | 70 | ||
XM_007646872.1 | XP_007645062.1 | 3 - 4 | 496 | 70 | ||
XM_007646873.1 | XP_007645063.1 | 2 - 3 | 590 | 70 | ||
XM_007646874.1 | XP_007645064.1 | 2 - 3 | 333 | 70 | ||
XM_007646875.1 | XP_007645065.1 | 2 - 3 | 332 | 70 | ||
GenBank mRNA | JI863339.1 | - | 2 - 3 | 335 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
