Assay Details
Gene Symbol: Rai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504210.2 | XP_003504258.1 | 1 | 1515 | 72 | |
XM_007644740.1 | XP_007642930.1 | 2 | 1365 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
