Assay Details
| Gene Symbol: Ndufb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003516009.2 | XP_003516057.2 | 1 | 216 | 66 | |
More Information
Set Membership: |
|
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufb9
Displaying...
| Gene Symbol: Ndufb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003516009.2 | XP_003516057.2 | 1 | 216 | 66 | |
Set Membership: |
|
Genomic Map