Assay Details
Gene Symbol: Mrpl46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514274.2 | XP_003514322.2 | 3 - 4 | 432 | 74 | |
GenBank mRNA | JP055849.1 | - | 3 - 4 | 432 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
