Assay Details
Gene Symbol: DRAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677912.3 | XP_005677969.1 | 6 - 7 | 720 | 86 | |
XM_005677913.3 | XP_005677970.1 | 7 - 8 | 771 | 86 | ||
XM_018045907.1 | XP_017901396.1 | 6 - 7 | 632 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
