Assay Details
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046969.1 | XP_017902458.1 | 4 - 5 | 1004 | 83 | |
XM_018046970.1 | XP_017902459.1 | 4 - 5 | 1016 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
