Assay Details
Gene Symbol: PTGER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314255.1 | NP_001301184.1 | 1 - 2 | 808 | 100 | |
GenBank mRNA | JX308617.1 | - | 1 - 2 | 808 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
