Assay Details
Gene Symbol: TMIGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693291.3 | XP_005693348.1 | 4 - 5 | 838 | 69 | |
GenBank mRNA | JP351172.2 | - | 2 - 3 | 186 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
