Assay Details
Gene Symbol: CD63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048119.1 | XP_017903608.1 | 2 - 3 | 422 | 71 | |
XM_018048120.1 | XP_017903609.1 | 2 - 3 | 204 | 71 | ||
XM_018048121.1 | XP_017903610.1 | 2 - 3 | 422 | 71 | ||
GenBank mRNA | JO363257.2 | - | 6 - 7 | 743 | 71 | |
JO579645.1 | - | 6 - 7 | 802 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
