Assay Details
Gene Symbol: DUSP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013975672.2 | XP_013831126.1 | 6 - 7 | 1068 | 71 | |
XM_018042603.1 | XP_017898092.1 | 7 - 8 | 1137 | 71 | ||
XM_018042604.1 | XP_017898093.1 | 7 - 8 | 1038 | 71 | ||
XM_018042605.1 | XP_017898094.1 | 5 - 6 | 635 | 71 | ||
XM_018042606.1 | XP_017898095.1 | 6 - 7 | 948 | 71 | ||
XM_018042608.1 | XP_017898097.1 | 5 - 6 | 878 | 71 | ||
GenBank mRNA | JT753913.1 | - | 5 - 6 | 1154 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
