Assay Details
Gene Symbol: LOC102184834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060680.1 | XP_017916169.1 | 31 - 32 | 4335 | 89 | |
XM_018060681.1 | XP_017916170.1 | 30 - 31 | 4278 | 89 | ||
XM_018060682.1 | XP_017916171.1 | 32 - 33 | 5639 | 89 | ||
XM_018060683.1 | XP_017916172.1 | 32 - 33 | 5639 | 89 | ||
XM_018060684.1 | XP_017916173.1 | 31 - 32 | 5582 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
