Assay Details
Gene Symbol: LOC102184834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060680.1 | XP_017916169.1 | 41 - 42 | 5420 | 86 | |
XM_018060681.1 | XP_017916170.1 | 40 - 41 | 5363 | 86 | ||
XM_018060682.1 | XP_017916171.1 | 42 - 43 | 6724 | 86 | ||
XM_018060683.1 | XP_017916172.1 | 42 - 43 | 6724 | 86 | ||
XM_018060684.1 | XP_017916173.1 | 41 - 42 | 6667 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
