Assay Details
Gene Symbol: LOC102186926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044705.1 | XP_017900194.1 | 4759 | 97 |
Gene Symbol: KIF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046471.1 | XP_017901960.1 | 17 - 18 | 2930 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
