Assay Details
Gene Symbol: LOC102172084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059551.1 | XP_017915040.1 | 33 - 34 | 5800 | 88 | |
XM_018059552.1 | XP_017915041.1 | 33 - 34 | 5796 | 88 | ||
XM_018059553.1 | XP_017915042.1 | 33 - 34 | 5800 | 88 | ||
XM_018059554.1 | XP_017915043.1 | 32 - 33 | 5737 | 88 | ||
XM_018059555.1 | XP_017915044.1 | 33 - 34 | 5677 | 88 | ||
XM_018059556.1 | XP_017915045.1 | 31 - 32 | 5611 | 88 | ||
XM_018059557.1 | XP_017915046.1 | 29 - 30 | 5230 | 88 | ||
XM_018059558.1 | XP_017915047.1 | 28 - 29 | 5167 | 88 | ||
XM_018059559.1 | XP_017915048.1 | 28 - 29 | 5122 | 88 | ||
XM_018059560.1 | XP_017915049.1 | 27 - 28 | 5058 | 88 | ||
GenBank mRNA | KA322253.1 | - | 4 - 5 | 919 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
