Assay Details
Gene Symbol: SARNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680393.3 | XP_005680450.1 | 7 - 8 | 528 | 73 | |
XM_018048118.1 | XP_017903607.1 | 8 - 9 | 669 | 73 | ||
GenBank mRNA | JO581508.1 | - | 7 - 8 | 673 | 73 | |
JT666863.1 | - | 7 - 8 | 654 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
