Assay Details
Gene Symbol: KRT39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972066.2 | XP_013827520.2 | 5 - 6 | 1076 | 73 | |
XM_013972067.2 | XP_013827521.2 | 5 - 6 | 1076 | 73 | ||
XM_013972068.2 | XP_013827522.2 | 5 - 6 | 1076 | 73 | ||
XM_013972069.2 | XP_013827523.2 | 6 - 7 | 867 | 73 | ||
XM_018065050.1 | XP_017920539.1 | 5 - 6 | 1076 | 73 | ||
GenBank mRNA | KA326566.1 | - | 2 - 3 | 2536 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
