Assay Details
Gene Symbol: ECM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002806481.4 | XP_002806527.2 | 8 - 9 | 2471 | 117 | |
XM_008992936.2 | XP_008991184.1 | 8 - 9 | 2540 | 117 | ||
XM_008992940.2 | XP_008991188.1 | 9 - 10 | 2592 | 117 | ||
XM_008992947.2 | XP_008991195.1 | 7 - 8 | 2345 | 117 | ||
XM_008992951.2 | XP_008991199.1 | 7 - 8 | 2277 | 117 | ||
XM_017971895.1 | XP_017827384.1 | 8 - 9 | 2566 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
