Assay Details
Gene Symbol: TRNAU1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750517.4 | XP_002750563.1 | 8 - 9 | 821 | 80 | |
XM_009000758.2 | XP_008999006.1 | 7 - 8 | 689 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
