Assay Details
Gene Symbol: TENT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002746762.3 | XP_002746808.1 | 2 - 3 | 897 | 98 | |
XM_002746763.3 | XP_002746809.1 | 2 - 3 | 1106 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
