Assay Details
Gene Symbol: IGSF21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002750355.4 | XP_002750401.3 | 6 - 7 | 1393 | 68 | |
GenBank mRNA | EF215414.1 | - | 1 - 2 | 180 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
