Assay Details
Gene Symbol: FRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002746529.3 | XP_002746575.1 | 4 - 5 | 514 | 58 | |
XM_008994415.2 | XP_008992663.1 | 5 - 6 | 566 | 58 | ||
XM_017971080.1 | XP_017826569.1 | 3 - 4 | 1513 | 58 | ||
XM_017971081.1 | XP_017826570.1 | 4 - 5 | 1083 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
