Assay Details
Gene Symbol: GNGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002751597.3 | XP_002751643.1 | 2 - 3 | 570 | 89 | |
XM_009002432.2 | XP_009000680.1 | 3 - 4 | 285 | 89 | ||
XM_009002436.1 | XP_009000684.1 | 2 - 3 | 286 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
