Assay Details
Gene Symbol: LOC100394535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756370.2 | XP_002756416.1 | 6 - 7 | 697 | 104 | |
XM_002756371.1 | XP_002756417.1 | 6 - 7 | 598 | 104 | ||
XM_009009931.1 | XP_009008179.1 | 6 - 7 | 722 | 104 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
