Assay Details
Gene Symbol: Apoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172915.1 | NP_001166386.1 | 2 - 3 | 206 | 86 | |
XM_013155371.2 | XP_013010825.1 | 4 - 5 | 366 | 86 | ||
GenBank mRNA | AF030571.1 | - | 2 - 3 | 206 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
