Assay Details
Gene Symbol: Ebp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172945.1 | NP_001166416.1 | 4 - 5 | 568 | 80 | |
GenBank mRNA | Z37985.1 | - | 4 - 5 | 568 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
