Assay Details
Gene Symbol: ckmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198801.1 | NP_942096.1 | 2 - 3 | 250 | 70 | |
XM_021470450.1 | XP_021326125.1 | 1 - 2 | 70 | |||
GenBank mRNA | BC048050.1 | - | 2 - 3 | 250 | 70 | |
BC165565.1 | - | 1 - 2 | 187 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
