Assay Details
| Gene Symbol: khsrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089428.2 | NP_001082897.2 | 4 - 5 | 413 | 103 | |
| XM_005160179.4 | XP_005160236.1 | 4 - 5 | 329 | 103 | ||
| GenBank mRNA | BC139701.1 | - | 4 - 5 | 414 | 103 | |
More Information
Set Membership: |
|

khsrp
Genomic Map