Assay Details
Gene Symbol: rpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200028.1 | NP_956322.1 | 369 | 71 | ||
GenBank mRNA | BC049055.1 | - | 369 | 71 | ||
BC062278.1 | - | 4 - 5 | 338 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
