Assay Details
Gene Symbol: MIR9051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128071.1 | 1 | 50 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-9045 | MIR9045 | eca-mir-9045 | -1.2kb | Sense | Chr. 1 - 109452091 - 109452225 on NCBI Build EquCab3.0 |
Downstream | OTU deubiquitinase 7A | OTUD7A | 71.5kb | Anti-sense | Chr. 1 - 109525100 - 109912253 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
