Assay Details
Gene Symbol: MIR488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032847.1 | 1 | 45 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | astrotactin 1 | ASTN1 | Sense | Chr. 5 - 12027052 - 12318396 on NCBI Build EquCab3.0 | ||
Downstream | BMP/retinoic acid inducible neural specific 2 | BRINP2 | 136.7kb | Anti-sense | Chr. 5 - 12323339 - 12427440 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
