Assay Details
Gene Symbol: MIR26A-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032954.1 | 1 | 33 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CTD small phosphatase like | CTDSPL | Sense | Chr. 16 - 48835858 - 48947420 on NCBI Build EquCab3.0 | ||
Downstream | integrin subunit alpha 9 | ITGA9 | 133.1kb | Sense | Chr. 16 - 48982030 - 49313604 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
