Assay Details
Gene Symbol: MIR9177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128255.2 | 1 | 79 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myosin light chain kinase | MYLK | Sense | Chr. 19 - 38284342 - 38542992 on NCBI Build EquCab3.0 | ||
Downstream | 3-hydroxyacyl-CoA dehydratase 2 | HACD2 | PTPLB | 219.2kb | Sense | Chr. 19 - 38556538 - 38653142 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
