Assay Details
Gene Symbol: PM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004945285.3 | XP_004945342.2 | 2 - 3 | 250 | 80 | |
XM_414903.6 | XP_414903.3 | 2 - 3 | 235 | 80 | ||
GenBank mRNA | CR386453.1 | - | 2 - 3 | 108 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
