Assay Details
| Gene Symbol: PM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004945285.3 | XP_004945342.2 | 7 - 8 | 736 | 65 | |
| XM_414903.6 | XP_414903.3 | 7 - 8 | 721 | 65 | ||
| GenBank mRNA | CR386453.1 | - | 7 - 8 | 594 | 65 | |
More Information
Set Membership: |
|

PM5
Genomic Map