Assay Details
Gene Symbol: COX7A2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277587.1 | NP_001264516.1 | 1 - 2 | 174 | 139 | |
XM_015283681.2 | XP_015139167.1 | 1 - 2 | 331 | 139 | ||
GenBank mRNA | BX932330.1 | - | 1 - 2 | 174 | 139 | |
CK613988.1 | - | 1 - 2 | 108 | 139 | ||
CN232533.1 | - | 1 - 2 | 121 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
