Assay Details
Gene Symbol: PPM1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031052.1 | NP_001026223.1 | 4 - 5 | 1195 | 81 | |
XM_015283686.2 | XP_015139172.2 | 4 - 5 | 1250 | 81 | ||
XM_015283687.2 | XP_015139173.2 | 6 - 7 | 1478 | 81 | ||
XM_015283688.2 | XP_015139174.2 | 5 - 6 | 1380 | 81 | ||
XM_015283689.2 | XP_015139175.2 | 4 - 5 | 1189 | 81 | ||
XM_015283690.2 | XP_015139176.2 | 3 - 4 | 1113 | 81 | ||
XM_015283692.2 | XP_015139178.2 | 4 - 5 | 1250 | 81 | ||
XM_015283693.2 | XP_015139179.2 | 4 - 5 | 1250 | 81 | ||
XM_025148676.1 | XP_025004444.1 | 4 - 5 | 1146 | 81 | ||
GenBank mRNA | AJ851753.1 | - | 4 - 5 | 1195 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
