Assay Details
Gene Symbol: VCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044664.1 | NP_001038129.1 | 3 - 4 | 459 | 60 | |
GenBank mRNA | AB195711.1 | - | 3 - 4 | 409 | 60 | |
AJ719285.1 | - | 3 - 4 | 459 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
