Assay Details
| Gene Symbol: LOC770450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233784.5 | XP_001233785.1 | 2 - 3 | 156 | 82 | |
More Information
Set Membership: |
|
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC770450| Gene Symbol: LOC770450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233784.5 | XP_001233785.1 | 2 - 3 | 156 | 82 | |
Set Membership: |
|