Assay Details
Gene Symbol: CREBRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153607.2 | NP_705835.2 | 6 - 7 | 1925 | 110 | |
XM_005265821.3 | XP_005265878.1 | 6 - 7 | 1942 | 110 | ||
XM_006714822.3 | XP_006714885.1 | 6 - 7 | 2115 | 110 | ||
XM_011534442.1 | XP_011532744.1 | 5 - 6 | 1728 | 110 | ||
GenBank mRNA | AK122675.1 | - | 3 - 4 | 871 | 110 | |
AK291677.1 | - | 6 - 7 | 1884 | 110 | ||
AK297424.1 | - | 6 - 7 | 2087 | 110 | ||
AY139008.1 | - | 6 - 7 | 1910 | 110 | ||
AY635785.1 | - | 4 - 5 | 1606 | 110 | ||
BC041709.1 | - | 6 - 7 | 1910 | 5761775 | 110 | |
BX647573.1 | - | 5 - 6 | 3586 | 110 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05000958.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
transport
response to unfolded protein
maternal behavior
negative regulation of glucocorticoid mediated signaling pathway
positive regulation of prolactin signaling pathway
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
transport
response to unfolded protein
maternal behavior
negative regulation of glucocorticoid mediated signaling pathway
positive regulation of prolactin signaling pathway
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05013937.hg.1 | TC05000958.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05013937.hg.1 | ENST00000296953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05013937.hg.1 | ENST00000393776.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05013937.hg.1 | ENST00000538538.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05013937.hg.1 | ENST00000540014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05013937.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05013932.hg.1 | TC05000958.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05013932.hg.1 | ENST00000296953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05013932.hg.1 | ENST00000393776.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05013932.hg.1 | ENST00000538538.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05013932.hg.1 | ENST00000540014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05013932.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05013936.hg.1 | TC05000958.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05013936.hg.1 | ENST00000296953.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05013936.hg.1 | ENST00000393776.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05013936.hg.1 | ENST00000538538.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05013936.hg.1 | ENST00000540014.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05013936.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000517882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000517882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000517882.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000523161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000523161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | ENST00000523161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500166406.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500166406.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500166406.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500166406.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500166406.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500166406.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500166406.hg.1 | TC0500009488.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500166405.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500166405.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500166405.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500166405.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500166405.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500166405.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500166405.hg.1 | TC0500009488.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000517882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000517882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000517882.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520420.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000522692.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000523161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000523161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | ENST00000523161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168393.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_001168394.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009488.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500166403.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500166403.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500166403.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500166403.hg.1 | ENST00000296953.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500166403.hg.1 | ENST00000520464.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500166403.hg.1 | NM_153607.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500166403.hg.1 | TC0500009488.hg.1 | Clariom D TC |
