Assay Details
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176814.4 | NP_789784.2 | 3 - 4 | 877 | 132 | |
XM_005250181.3 | XP_005250238.1 | 3 - 4 | 951 | 132 | ||
XM_006715867.3 | XP_006715930.1 | 3 - 4 | 956 | 132 | ||
XM_006715868.3 | XP_006715931.1 | 3 - 4 | 951 | 132 | ||
XM_011515859.2 | XP_011514161.1 | 3 - 4 | 672 | 132 | ||
GenBank mRNA | AF218032.1 | - | 3 - 4 | 847 | 132 | |
AK297946.1 | - | 1 - 2 | 65 | 132 | ||
AK309674.1 | - | 3 - 4 | 871 | 132 | ||
BC047641.1 | - | 3 - 4 | 896 | 132 | ||
BC110623.1 | - | 3 - 4 | 422 | 40036036 | 132 | |
BC110624.1 | - | 3 - 4 | 417 | 40036038 | 132 | |
DQ655936.2 | - | 3 - 4 | 268 | 132 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07001835.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK297946.1,BC047641.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000439506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000439506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000439506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000439506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000439506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07028433.hg.1 | TC07001835.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07028433.hg.1 | ENST00000265827.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07028433.hg.1 | ENST00000393312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07028433.hg.1 | ENST00000393313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07028433.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000439506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000439506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000439506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000439506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000439506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013615.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000439506.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000265827.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000393312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000393313.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000434602.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000436992.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000439506.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | ENST00000619291.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | NM_176814.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700201860.hg.1 | TC0700013615.hg.1 | Clariom D TC |
